Please login to your online account to display your discounted pricing

2-(2-chlorophenoxy)ethylamine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(2-Chlorophenoxy)ethylamine
CAS 26378-53-0
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H10ClNO
Linear Formula ClC6H4O(CH2)2NH2
Formula Weight 171.63
MDL Number MFCD00125294
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RF03300CB

  SDS
fisher scientific company
RF03300CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.63
Quantity 250mg
Assay 97%
CAS Min % 97.0
Linear Formula ClC6H4O(CH2)2NH2
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 26378-53-0
Chemical Name or Material 2-(2-Chlorophenoxy)ethylamine
Molecular Formula C8H10ClNO
MDL Number MFCD00125294
Health Hazard 1 GHS Signal Word: Danger
Packaging Amber Glass Bottle
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.