Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Chlorophenoxy)ethylamine, 95%

Chemical Identifiers

Chemical Name or Material 2-(2-Chlorophenoxy)ethylamine
CAS 26378-53-0
Molecular Formula C8H10ClNO
Formula Weight 171.63
MDL Number MFCD00125294
UN Number UN2735
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5070403

  SDS
alfa aesar™
H5070403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH50704MD

  SDS
alfa aesar™
H50704MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.63
Chemical Name or Material 2-(2-Chlorophenoxy)ethylamine
Assay 95%
Molecular Formula C8H10ClNO
MDL Number MFCD00125294
CAS 26378-53-0
UN Number UN2735
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Boiling Point 150° to 153°C (18mmHg)
Melting Point 39° to 40°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.