Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Chlorophenoxy)ethanol, 98%

Chemical Identifiers

Chemical Name or Material 2-(2-Chlorophenoxy)ethanol
CAS 15480-00-9
Molecular Formula C8H9ClO2
Formula Weight 172.61
MDL Number MFCD00014034
UN Number UN2810
Beilstein 1945576
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0436822

 
alfa aesar™
L0436822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0436814

 
alfa aesar™
L0436814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.61
Chemical Name or Material 2-(2-Chlorophenoxy)ethanol
Molecular Formula C8H9ClO2
MDL Number MFCD00014034
CAS 15480-00-9
TSCA TSCA
Assay 98%
UN Number UN2810
Beilstein 1945576
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Flash Point >110°C (230°F)
Boiling Point 114° to 116°C (1mmHg)
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.