Please login to your online account to display your discounted pricing

2-(2-chlorophenoxy)ethanimidamide hydrochloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(2-Chlorophenoxy)ethanimidamide hydrochloride
CAS 58403-03-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H9ClN2O·ClH
Linear Formula ClC6H4OCH2C(=NH)NH2·HCl
Formula Weight 221.09
MDL Number MFCD00173805
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB05621DA

 
fisher scientific company
SPB05621DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SPB05621EA

 
fisher scientific company
SPB05621EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221.09
Quantity 1g
Assay 97%
CAS Min % 97.0
Linear Formula ClC6H4OCH2C(=NH)NH2·HCl
CAS 58403-03-5
Packaging Amber glass bottle
Chemical Name or Material 2-(2-Chlorophenoxy)ethanimidamide hydrochloride
Assay Percent Range 97%
Molecular Formula C8H9ClN2O·ClH
MDL Number MFCD00173805
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.