Please login to your online account to display your discounted pricing

2-(2-chlorophenoxy)ethanimidamide hydrochloride, 97%, Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SPB05621EA

 View more versions of this product

Catalog No. SPB05621EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 221.09
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 2-(2-Chlorophenoxy)ethanimidamide hydrochloride
Assay Percent Range 97%
Linear Formula ClC6H4OCH2C(=NH)NH2·HCl
CAS 58403-03-5
Molecular Formula C8H9ClN2O·ClH
MDL Number MFCD00173805
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.

Specifications

Chemical Name or Material 2-(2-Chlorophenoxy)ethanimidamide hydrochloride
CAS 58403-03-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H9ClN2O·ClH
Linear Formula ClC6H4OCH2C(=NH)NH2·HCl
Formula Weight 221.09
MDL Number MFCD00173805
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.