Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Chlorophenoxy)acetamide, 97%

$43.08 - $601.20

Chemical Identifiers

Chemical Name or Material 2-(2-Chlorophenoxy)acetamide
CAS 35368-68-4
Molecular Formula C8H8ClNO2
Formula Weight 185.61
MDL Number MFCD00173819
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2522303

  SDS
alfa aesar™
B2522303
1g Each for $43.08

AAB2522306

  SDS
alfa aesar™
B2522306
5g Each for $150.68

AAB2522314

  SDS
alfa aesar™
B2522314
25g Each for $601.20
Description & Specifications

Specifications

Formula Weight 185.61
Assay 97%
CAS 35368-68-4
Quantity 1g
Chemical Name or Material 2-(2-Chlorophenoxy)acetamide
Molecular Formula C8H8ClNO2
MDL Number MFCD00173819
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.