missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(2-Chloroethoxy)ethanol, 99%, ACROS Organics™
$62.10 - $62.10
Chemical Identifiers
CAS | 628-89-7 |
---|---|
Molecular Formula | C4H9ClO2 |
Molecular Weight (g/mol) | 124.57 |
MDL Number | MFCD00002870 |
InChI Key | LECMBPWEOVZHKN-UHFFFAOYSA-N |
Synonym | 2-2-chloroethoxy ethan-1-ol, 2-2-chloroethoxy ethanol, 2-chloroethyl 2-hydroxyethyl ether, diethylene glycol monochlorohydrin, diglycol chlorhydrin, diglycol chlorohydrin, ethanol, 2-2-chloroethoxy, monochloropolyoxyethylene, polyethylene glycol chloride 210, unii-r2up818l4i |
PubChem CID | 12361 |
IUPAC Name | 2-(2-chloroethoxy)ethanol |
SMILES | C(COCCCl)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC172260500
|
Thermo Fisher Scientific
172260500 |
50mL | Glass bottle |
Each for $62.10
|
|
AC172262500
|
Thermo Fisher Scientific
172262500 |
250mL | Glass bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
628-89-7 | |
124.57 | |
LECMBPWEOVZHKN-UHFFFAOYSA-N | |
12361 | |
C(COCCCl)O |
C4H9ClO2 | |
MFCD00002870 | |
2-2-chloroethoxy ethan-1-ol, 2-2-chloroethoxy ethanol, 2-chloroethyl 2-hydroxyethyl ether, diethylene glycol monochlorohydrin, diglycol chlorhydrin, diglycol chlorohydrin, ethanol, 2-2-chloroethoxy, monochloropolyoxyethylene, polyethylene glycol chloride 210, unii-r2up818l4i | |
2-(2-chloroethoxy)ethanol |
Specifications
1.1800g/mL | |
Authentic | |
1.4519 to 1.4539 | |
1% max. (K.F.) | |
Undesignated | |
628-89-7 | |
100.0 | |
C4H9ClO2 | |
MFCD00002870 | |
2-2-chloroethoxy ethan-1-ol, 2-2-chloroethoxy ethanol, 2-chloroethyl 2-hydroxyethyl ether, diethylene glycol monochlorohydrin, diglycol chlorhydrin, diglycol chlorohydrin, ethanol, 2-2-chloroethoxy, monochloropolyoxyethylene, polyethylene glycol chloride 210, unii-r2up818l4i | |
LECMBPWEOVZHKN-UHFFFAOYSA-N | |
2-(2-chloroethoxy)ethanol | |
12361 | |
Liquid | |
2-(2-Chloroethoxy)ethanol, 99% |
90°C | |
Glass bottle | |
1.18 | |
180°C to 185°C | |
50mL | |
98.5 | |
98.5% min. (GC) | |
ClCH2CH2OCH2CH2OH | |
01, 467 | |
Solubility in water: very soluble. Other solubilities: miscible with ethyl alcohol and diethyl ether | |
C(COCCCl)O | |
124.57 | |
124.57 | |
99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 211-059-9
RTECSNumber : KK1350000
TSCA : TSCA