Learn More
2-(2-Chloroethoxy)ethanol, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 172262500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-(2-Chloroethoxy)ethanol | |
628-89-7 | |
100.0 | |
180°C to 185°C | |
Authentic | |
Glass bottle | |
1.4519 to 1.4539 | |
MFCD00002870 | |
01, 467 | |
2-2-chloroethoxy ethanol, ethanol, 2-2-chloroethoxy, diglycol chlorohydrin, diglycol chlorhydrin, 2-chloroethyl 2-hydroxyethyl ether, monochloropolyoxyethylene, 2-2-chloroethoxy ethan-1-ol, unii-r2up818l4i, diethylene glycol monochlorohydrin, polyethylene glycol chloride 210 | |
LECMBPWEOVZHKN-UHFFFAOYSA-N | |
2-(2-chloroethoxy)ethanol | |
12361 | |
99% |
99% | |
98.5 | |
1.1800g/mL | |
90°C | |
98.5% min. (GC) | |
C4H9ClO2 | |
ClCH2CH2OCH2CH2OH | |
250mL | |
1.18 | |
Solubility in water: very soluble. Other solubilities: miscible with ethyl alcohol and diethyl ether | |
OCCOCCCl | |
124.56 | |
124.57 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 211-059-9
RTECSNumber : KK1350000
TSCA : TSCA
RUO â Research Use Only