missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[(2-chloro-6-fluorobenzyl)thio]ethylamine, 97%, Maybridge

Chemical Identifiers

CAS 175136-76-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H11ClFNS
Formula Weight 219.71
MDL Number MFCD00052683
Chemical Name or Material 2-[(2-chloro-6-fluorobenzyl)thio]ethylamine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CD07751EA SDS Fisher Scientific Company
CD07751EA
Amber Glass Bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

175136-76-2
100.0
C9H11ClFNS
MFCD00052683
97%
96.5
97%
219.71
2-[(2-chloro-6-fluorobenzyl)thio]ethylamine

Specifications

219.71
100.0
MFCD00052683
Amber Glass Bottle
97%
97%
10g
C9H11ClFNS
96.5
175136-76-2
2-[(2-chloro-6-fluorobenzyl)thio]ethylamine

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger