missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Carboxyvinyl)benzeneboronic acid, 98%

$95.92 - $339.27

Chemical Identifiers

CAS 374105-86-9
Molecular Formula C9H9BO4
Molecular Weight (g/mol) 191.977
MDL Number MFCD02093067
InChI Key OPFDGNUJQALQFL-AATRIKPKSA-N
Synonym 2-2-carboxyvinyl benzeneboronic acid, 2-boronocinnamic acid, 3-2-boronophenyl acrylic acid, e-3-2-boronophenyl prop-2-enoic acid, e-3-2-boronophenyl acrylic acid, 2e-3-2-dihydroxyboranyl phenyl prop-2-enoic acid, 2-2-carboxyvinylbenzene boronic acid, 2-2-carboxyvinyl phenylboronic acid, 2-2-carboxy-vinyl-phenylboronic acid, e-3-2-boronophenyl-2-propenoic acid
PubChem CID 6158001
IUPAC Name (E)-3-(2-boronophenyl)prop-2-enoic acid
SMILES B(C1=CC=CC=C1C=CC(=O)O)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL16368MD View Documents Alfa Aesar™
L16368MD
250mg Each for $339.27
 
AAL16368MF View Documents Alfa Aesar™
L16368MF
50mg Each for $95.92
 
Specifications

Chemical Identifiers

374105-86-9
191.977
OPFDGNUJQALQFL-AATRIKPKSA-N
6158001
B(C1=CC=CC=C1C=CC(=O)O)(O)O
C9H9BO4
MFCD02093067
2-2-carboxyvinyl benzeneboronic acid, 2-boronocinnamic acid, 3-2-boronophenyl acrylic acid, e-3-2-boronophenyl prop-2-enoic acid, e-3-2-boronophenyl acrylic acid, 2e-3-2-dihydroxyboranyl phenyl prop-2-enoic acid, 2-2-carboxyvinylbenzene boronic acid, 2-2-carboxyvinyl phenylboronic acid, 2-2-carboxy-vinyl-phenylboronic acid, e-3-2-boronophenyl-2-propenoic acid
(E)-3-(2-boronophenyl)prop-2-enoic acid

Specifications

170°C to 172°C
374105-86-9
MFCD02093067
OPFDGNUJQALQFL-AATRIKPKSA-N
(E)-3-(2-boronophenyl)prop-2-enoic acid
6158001
98%
250mg
C9H9BO4
2-2-carboxyvinyl benzeneboronic acid, 2-boronocinnamic acid, 3-2-boronophenyl acrylic acid, e-3-2-boronophenyl prop-2-enoic acid, e-3-2-boronophenyl acrylic acid, 2e-3-2-dihydroxyboranyl phenyl prop-2-enoic acid, 2-2-carboxyvinylbenzene boronic acid, 2-2-carboxyvinyl phenylboronic acid, 2-2-carboxy-vinyl-phenylboronic acid, e-3-2-boronophenyl-2-propenoic acid
B(C1=CC=CC=C1C=CC(=O)O)(O)O
191.977
191.98
2-(2-Carboxyvinyl)benzeneboronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning