missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(2-Butoxyethoxy)ethanol, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC149710100

 View more versions of this product

Catalog No. AC149710100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-(2-Butoxyethoxy)ethanol
100°C
1.4310 to 1.4330
6.16 mPa.s (20°C)
-68.0°C
99+%
99.0
99% min. (GC)
CH3(CH2)3OCH2CH2OCH2CH2OH
15,3133
Solubility in water: soluble. Other solubilities: miscible with oils, miscible with most common organic solvents
CCCCOCCOCCO
162.23
162.23
0.9550g/mL
Plastic drum
0.955
231.0°C
10L
112-34-5
100.0
C8H18O3
01,IV,2394
2-2-butoxyethoxy ethanol, butoxydiglycol, butoxyethoxyethanol, butyl carbitol, butyl diglycol, butyl digol, butyldiglycol, diethylene glycol butyl ether, diethylene glycol monobutyl ether, ethanol, 2-2-butoxyethoxy
OAYXUHPQHDHDDZ-UHFFFAOYSA-N
2-(2-butoxyethoxy)ethanol
8177
99+%

Chemical Identifiers

112-34-5
162.23
2-2-butoxyethoxy ethanol, butoxydiglycol, butoxyethoxyethanol, butyl carbitol, butyl diglycol, butyl digol, butyldiglycol, diethylene glycol butyl ether, diethylene glycol monobutyl ether, ethanol, 2-2-butoxyethoxy
2-(2-butoxyethoxy)ethanol
C8H18O3
OAYXUHPQHDHDDZ-UHFFFAOYSA-N
8177
CCCCOCCOCCO
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-961-6

SDS Documents