missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(2-Bromophenyl)ethylamine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6073904

 View more versions of this product

Catalog No. AAH6073904



Description

Description

2-(2-Bromophenyl)ethylamine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Bromophenyl)ethylamine is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

2-(2-Bromophenyl)ethylamine
1.396
C8H10BrN
MFCD01529874
UN2735
2-bromophenethylamine, 2-2-bromophenyl ethanamine, 2-2-bromophenyl ethylamine, 2-2-bromophenyl ethan-1-amine, benzeneethanamine, 2-bromo, 2-bromo phenethylamin, 2-2-bromopheneyl ethylamine, 1-amino-2-bromophenyl ethane, 2-bromo phenethylamine, 2-bromo-phenethylamine
C1=CC=C(C(=C1)CCN)Br
200.079
200.08
65185-58-2
253°C
1.577
2 g
Air Sensitive
ITRNQMJXZUWZQL-UHFFFAOYSA-N
2-(2-bromophenyl)ethanamine
2734091
97%

Chemical Identifiers

65185-58-2
200.079
ITRNQMJXZUWZQL-UHFFFAOYSA-N
2734091
C1=CC=C(C(=C1)CCN)Br
C8H10BrN
MFCD01529874
2-bromophenethylamine, 2-2-bromophenyl ethanamine, 2-2-bromophenyl ethylamine, 2-2-bromophenyl ethan-1-amine, benzeneethanamine, 2-bromo, 2-bromo phenethylamin, 2-2-bromopheneyl ethylamine, 1-amino-2-bromophenyl ethane, 2-bromo phenethylamine, 2-bromo-phenethylamine
2-(2-bromophenyl)ethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only