missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(2-Bromophenyl)-1,3-dioxolane 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC393240010

 View more versions of this product

Catalog No. AC393240010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
Liquid
Clear colorless to pale yellow
1.5670 to 1.5690 (20°C, 589nm)
2-Bromobenzaldehyde ethylene acetal
C9H9BrO2
97.5
98%
229.07
1g
100.0
Glass bottle
2-(2-Bromophenyl)-1, 3-dioxolane
34824-58-3
MFCD00155124
Authentic

Chemical Identifiers

34824-58-3
100.0
C9H9BrO2
MFCD00155124
2-(2-Bromophenyl)-1, 3-dioxolane
97.5
98%
229.07
2-Bromobenzaldehyde ethylene acetal
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.