Learn More
2-(2-Bromoethyl)-1,3-dioxolane, 95%, stab. with silver, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1144118
Description
2-(2-Bromoethyl)-1,3-dioxolane is used as a pharmaceutical intermediate, dioxolanes, acetals, ketals, building blocks, chemical synthesis, organic building blocks and oxygen compounds. 1,1-(Ethylenedioxy)-3-bromopropane is used in the synthesis of EGFR inhibitors. Also used in the synthesis of orally active agents of cancer and cell proliferation with quinoline substructures. It is also used as flavouring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(2-Bromoethyl)-1,3-dioxolane is used as a pharmaceutical intermediate, dioxolanes, acetals, ketals, building blocks, chemical synthesis, organic building blocks and oxygen compounds. 1,1-(Ethylenedioxy)-3-bromopropane is used in the synthesis of EGFR inhibitors. Also used in the synthesis of orally active agents of cancer and cell proliferation with quinoline substructures. It is also used as flavouring agent.
Solubility
It is immiscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Light sensitive. Store under inert gas.
Specifications
2- (2-Bromoethyl)-1,3-dioxolane | |
18742-02-4 | |
68°C to 70°C (8 mmHg) | |
C5H9BrO2 | |
MFCD00003216 | |
UN2810 | |
Light sensitive | |
It is immiscible with water. | |
BrCCC1OCCO1 | |
181.03 | |
181.04 |
Stabilized with silver | |
1.512 | |
65°C (149°F) | |
1.479 | |
50 g | |
103516 | |
2-2-bromoethyl-1,3-dioxolane, 1,3-dioxolane, 2-2-bromoethyl, bromoethyl-1,3-dioxolane, acmc-1bol0, 2-2-bromoethyl-dioxolane, ksc496m8b, 2-bromoethyl-1,3-dioxolane, 2 2-bromoethyl-1,3-dioxolane, 2-2-bromoethyl-1,3-dioxolan | |
GGZQLTVZPOGLCC-UHFFFAOYSA-N | |
2-(2-bromoethyl)-1,3-dioxolane | |
87776 | |
95% |
Chemical Identifiers
18742-02-4 | |
181.03 | |
GGZQLTVZPOGLCC-UHFFFAOYSA-N | |
87776 | |
BrCCC1OCCO1 |
C5H9BrO2 | |
MFCD00003216 | |
2-2-bromoethyl-1,3-dioxolane, 1,3-dioxolane, 2-2-bromoethyl, bromoethyl-1,3-dioxolane, acmc-1bol0, 2-2-bromoethyl-dioxolane, ksc496m8b, 2-bromoethyl-1,3-dioxolane, 2 2-bromoethyl-1,3-dioxolane, 2-2-bromoethyl-1,3-dioxolan | |
2-(2-bromoethyl)-1,3-dioxolane |
Safety and Handling
GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 242-551-1
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only