missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(2-Bromoethyl)-1,3-dioxane 97.0+%, TCI America™
$255.49 - $255.49
Chemical Identifiers
CAS | 33884-43-4 |
---|---|
Molecular Formula | C6H11BrO2 |
Molecular Weight (g/mol) | 195.06 |
MDL Number | MFCD00006567 |
InChI Key | KXMZOKKPQZRPRN-UHFFFAOYNA-N |
Synonym | 2-2-bromoethyl-1,3-dioxane, 1,3-dioxane, 2-2-bromoethyl, 3-bromopropionaldehyde trimethylene acetal, 2 2-bromoethyl-1,3-dioxane, 2-2-bromoethyl-1,3 dioxane, bromoethyl-1,3-dioxane, acmc-209i3o, 2-2-bromoetyl 1,3-dioxane, wmdhqehpovoeog-uhfffaoysa, 2-2-bromoethyl 1,3-dioxane |
PubChem CID | 520656 |
IUPAC Name | 2-(1-bromoethyl)-1,3-dioxane |
SMILES | CC(Br)C1OCCCO1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B118225G
|
TCI America
B118225G |
25 g |
Each for $255.49
|
|
|||||
Chemical Identifiers
33884-43-4 | |
195.06 | |
KXMZOKKPQZRPRN-UHFFFAOYNA-N | |
520656 | |
CC(Br)C1OCCCO1 |
C6H11BrO2 | |
MFCD00006567 | |
2-2-bromoethyl-1,3-dioxane, 1,3-dioxane, 2-2-bromoethyl, 3-bromopropionaldehyde trimethylene acetal, 2 2-bromoethyl-1,3-dioxane, 2-2-bromoethyl-1,3 dioxane, bromoethyl-1,3-dioxane, acmc-209i3o, 2-2-bromoetyl 1,3-dioxane, wmdhqehpovoeog-uhfffaoysa, 2-2-bromoethyl 1,3-dioxane | |
2-(1-bromoethyl)-1,3-dioxane |
Specifications
33884-43-4 | |
70°C | |
MFCD00006567 | |
2-2-bromoethyl-1,3-dioxane, 1,3-dioxane, 2-2-bromoethyl, 3-bromopropionaldehyde trimethylene acetal, 2 2-bromoethyl-1,3-dioxane, 2-2-bromoethyl-1,3 dioxane, bromoethyl-1,3-dioxane, acmc-209i3o, 2-2-bromoetyl 1,3-dioxane, wmdhqehpovoeog-uhfffaoysa, 2-2-bromoethyl 1,3-dioxane | |
CC(Br)C1OCCCO1 | |
195.06 | |
195.06 | |
Liquid |
Yellow | |
C6H11BrO2 | |
25 g | |
KXMZOKKPQZRPRN-UHFFFAOYNA-N | |
2-(1-bromoethyl)-1,3-dioxane | |
520656 | |
≥97.0% (GC) | |
2-(2-Bromoethyl)-1,3-dioxane |
Safety and Handling
TSCA : Yes