Please login to your online account to display your discounted pricing

2-(2-Bromoethoxy)tetrahydro-2H-pyran, 96%, ACROS Organics™


Quantity 25mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC394930250

 View more versions of this product

Catalog No. AC394930250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 209.08
Quantity 25mL
Density 1.384g/mL
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C7H13BrO2
CAS 17739-45-6
Chemical Name or Material 2-(2-Bromoethoxy)tetrahydro-2H-pyran
MDL Number MFCD01321310
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 2-(2-Bromoethoxy)tetrahydro-2H-pyran
CAS 17739-45-6
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C7H13BrO2
Formula Weight 209.08
MDL Number MFCD01321310
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents