Please login to your online account to display your discounted pricing

2-(2-Bromoacetyl)thiazole, 97%, Acros Organics


Quantity 5g

Manufacturer: thermo fisher scientific  AC451470050

 View more versions of this product

Catalog No. AC451470050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 2-(2-Bromoacetyl)thiazole
CAS 3292-77-1
Assay Percent Range 97%
Molecular Formula C5H4BrNOS
Formula Weight 206.06
Quantity 5g

Specifications

Chemical Name or Material 2-(2-Bromoacetyl)thiazole
Assay Percent Range 97%
Molecular Formula C5H4BrNOS
Formula Weight 206.06