missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(2-Bromoacetyl)thiazole, 97%, Acros Organics

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C5H4BrNOS
Formula Weight 206.06
Chemical Name or Material 2-(2-Bromoacetyl)thiazole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC451470010 SDS Thermo Fisher Scientific
AC451470010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC451470050 SDS Thermo Fisher Scientific
AC451470050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
C5H4BrNOS
206.06
2-(2-Bromoacetyl)thiazole

Specifications

3292-77-1
C5H4BrNOS
1g
97%
206.06
2-(2-Bromoacetyl)thiazole

Safety and Handling

Recommended Storage : 4°C