Please login to your online account to display your discounted pricing

2,2-Bithiophene-5-carboxylic acid, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC14601CB

 View more versions of this product

Catalog No. CC14601CB

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 210.28
Quantity 250mg
Assay 97%
Chemical Name or Material 2, 2'-Bithiophene-5-carboxylic acid
Assay Percent Range 97%
Molecular Formula C9H6O2S2
CAS 2060-55-1
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material 2, 2'-Bithiophene-5-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H6O2S2
Formula Weight 210.28
Accessories

GHS Signal Word: Warning

SDS & Safety Documents