missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2'-Bithiophene-5-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3798903

 View more versions of this product

Catalog No. AAH3798903



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,2'-Bithiophene-5-boronic acid pinacol ester
35°C to 38°C
C14H17BO2S2
1 g
HPOQARMSOPOZMW-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-(5-thiophen-2-ylthiophen-2-yl)-1,3,2-dioxaborolane
3592790
98%
479719-88-5
Blue-Green to White
MFCD04039974
2,2'-bithiophene-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,2'-bithiophene, 2-2,2'-bithiophen-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-thiophen-2-yl thiophen-2-yl-1,3,2-dioxaborolane, acmc-209kb7, 4,4,5,5-tetramethyl-2-5-thiophen-2-ylthiophen-2-yl-1,3,2-dioxaborolane, 2,2'-bithiophene-5'-boronic acid pinacol ester
CC1(C)OB(OC1(C)C)C1=CC=C(S1)C1=CC=CS1
292.22
292.2
Fused Solid

Chemical Identifiers

479719-88-5
292.22
HPOQARMSOPOZMW-UHFFFAOYSA-N
3592790
CC1(C)OB(OC1(C)C)C1=CC=C(S1)C1=CC=CS1
C14H17BO2S2
MFCD04039974
2,2'-bithiophene-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,2'-bithiophene, 2-2,2'-bithiophen-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-thiophen-2-yl thiophen-2-yl-1,3,2-dioxaborolane, acmc-209kb7, 4,4,5,5-tetramethyl-2-5-thiophen-2-ylthiophen-2-yl-1,3,2-dioxaborolane, 2,2'-bithiophene-5'-boronic acid pinacol ester
4,4,5,5-tetramethyl-2-(5-thiophen-2-ylthiophen-2-yl)-1,3,2-dioxaborolane
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only