missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2′-Bis(trifluoromethyl)benzidine, TCI America™

$68.75 - $630.98

Chemical Identifiers

CAS 341-58-2
Molecular Formula C14H10F6N2
Molecular Weight (g/mol) 320.238
MDL Number MFCD00190155
InChI Key NVKGJHAQGWCWDI-UHFFFAOYSA-N
Synonym 2,2'-bis trifluoromethyl benzidine, 2,2'-bis trifluoromethyl-1,1'-biphenyl-4,4'-diamine, 4-4-amino-2-trifluoromethyl phenyl-3-trifluoromethyl aniline, 2,2'-bis trifluoromethyl-4,4'-diaminobiphenyl, 2,2'-bis-trifluoromethyl-biphenyl-4,4'-diamine, tfmb, 2,2'-bis trifluoromethyl-4,4'-biphenyldiamine, 4,4'-diamino-2,2'-bis trifluoromethyl biphenyl, 1,1'-biphenyl-4,4'-diamine, 2,2'-bis trifluoromethyl, 4-4-azanyl-2-trifluoromethyl phenyl-3-trifluoromethyl aniline
PubChem CID 629349
IUPAC Name 4-[4-amino-2-(trifluoromethyl)phenyl]-3-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1N)C(F)(F)F)C2=C(C=C(C=C2)N)C(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B17111G View Documents TCI America
B17111G
1g Each for $68.75
 
B17115G View Documents TCI America
B17115G
5g Each for $209.31
 
B171125G View Documents TCI America
B171125G
25g Each for $630.98
 
Specifications

Chemical Identifiers

341-58-2
320.238
NVKGJHAQGWCWDI-UHFFFAOYSA-N
629349
C1=CC(=C(C=C1N)C(F)(F)F)C2=C(C=C(C=C2)N)C(F)(F)F
C14H10F6N2
MFCD00190155
2,2'-bis trifluoromethyl benzidine, 2,2'-bis trifluoromethyl-1,1'-biphenyl-4,4'-diamine, 4-4-amino-2-trifluoromethyl phenyl-3-trifluoromethyl aniline, 2,2'-bis trifluoromethyl-4,4'-diaminobiphenyl, 2,2'-bis-trifluoromethyl-biphenyl-4,4'-diamine, tfmb, 2,2'-bis trifluoromethyl-4,4'-biphenyldiamine, 4,4'-diamino-2,2'-bis trifluoromethyl biphenyl, 1,1'-biphenyl-4,4'-diamine, 2,2'-bis trifluoromethyl, 4-4-azanyl-2-trifluoromethyl phenyl-3-trifluoromethyl aniline
4-[4-amino-2-(trifluoromethyl)phenyl]-3-(trifluoromethyl)aniline

Specifications

Undesignated
1g
C14H10F6N2
2,2'-bis trifluoromethyl benzidine, 2,2'-bis trifluoromethyl-1,1'-biphenyl-4,4'-diamine, 4-4-amino-2-trifluoromethyl phenyl-3-trifluoromethyl aniline, 2,2'-bis trifluoromethyl-4,4'-diaminobiphenyl, 2,2'-bis-trifluoromethyl-biphenyl-4,4'-diamine, tfmb, 2,2'-bis trifluoromethyl-4,4'-biphenyldiamine, 4,4'-diamino-2,2'-bis trifluoromethyl biphenyl, 1,1'-biphenyl-4,4'-diamine, 2,2'-bis trifluoromethyl, 4-4-azanyl-2-trifluoromethyl phenyl-3-trifluoromethyl aniline
C1=CC(=C(C=C1N)C(F)(F)F)C2=C(C=C(C=C2)N)C(F)(F)F
320.238
320.24
≥98.0% (GC,T)
183°C
341-58-2
MFCD00190155
NVKGJHAQGWCWDI-UHFFFAOYSA-N
4-[4-amino-2-(trifluoromethyl)phenyl]-3-(trifluoromethyl)aniline
629349
Crystal-Powder at 20°C
2,2′-Bis(trifluoromethyl)benzidine
Safety and Handling

Safety and Handling

TSCA : No