Learn More
2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 168870025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2,2-Bis(hydroxymethyl)-2,2′,2′′-nitrilotriethanol | |
6976-37-0 | |
(0.1M in H2O) | |
0.2% max. (80°C, 5 hrs) | |
99+% | |
C8H19NO5 | |
MFCD00002853 | |
bis-tris, bistris, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis-tris buffer, 1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, unii-q1xc3631cp, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl iminotris hydroxymethyl methane | |
OWMVSZAMULFTJU-UHFFFAOYSA-N | |
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol | |
81462 | |
209.24 |
99+% | |
99.0 | |
100.0 | |
Authentic | |
Plastic bottle | |
(HOCH2CH2)2NC(CH2OH)3 | |
2.5kg | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
C(CO)N(CCO)C(CO)(CO)CO | |
209.24 | |
CHEBI:41250 | |
99+% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 230-237-7
TSCA : TSCA
RUO â Research Use Only