missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 168871000

 View more versions of this product

Catalog No. AC168871000



Description

Description

Specifications

Specifications

(0.1M in H2O)
2,2-Bis(hydroxymethyl)-2,2′,2′′-nitrilotriethanol
0.2% max. (80°C, 5 hrs)
0.1% max.
Colorless to Yellow
100g
6976-37-0
100.0
C8H19NO5
MFCD00002853
Solubility in water: soluble. Other solubilities: soluble in alcohol
C(CO)N(CCO)C(CO)(CO)CO
209.24
CHEBI:41250
Liquid
(21% aq. soln.) clear, colorless
Authentic
Plastic bottle
1% Max. (K.F.)
103.0°C to 104.0°C
99+%
99.0
99+%
(HOCH2CH2)2NC(CH2OH)3
1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis 2-hydroxyethyl iminotris hydroxymethyl methane, bis-tris, bis-tris buffer, bistris, unii-q1xc3631cp
OWMVSZAMULFTJU-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
81462
209.24
99+%

Chemical Identifiers

6976-37-0
209.24
OWMVSZAMULFTJU-UHFFFAOYSA-N
81462
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
C8H19NO5
MFCD00002853
1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis 2-hydroxyethyl iminotris hydroxymethyl methane, bis-tris, bis-tris buffer, bistris, unii-q1xc3631cp
CHEBI:41250
C(CO)N(CCO)C(CO)(CO)CO
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 230-237-7

TSCA : TSCA

SDS Documents