missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, 98%, Thermo Scientific™

Chemical Identifiers

CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.24
MDL Number MFCD00002853
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
Synonym 1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis 2-hydroxyethyl iminotris hydroxymethyl methane, bis-tris, bis-tris buffer, bistris, unii-q1xc3631cp
PubChem CID 81462
ChEBI CHEBI:41250
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CO)N(CCO)C(CO)(CO)CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC163821000
SDS View Documents Promotion Details
Thermo Fisher Scientific
163821000
100g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC163825000
SDS View Documents Promotion Details
Thermo Fisher Scientific
163825000
500g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC163820025
SDS View Documents Promotion Details
Thermo Fisher Scientific
163820025
2.5kg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6976-37-0
209.24
OWMVSZAMULFTJU-UHFFFAOYSA-N
81462
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
C8H19NO5
MFCD00002853
1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis 2-hydroxyethyl iminotris hydroxymethyl methane, bis-tris, bis-tris buffer, bistris, unii-q1xc3631cp
CHEBI:41250
C(CO)N(CCO)C(CO)(CO)CO

Specifications

6976-37-0
(0.1M in H2O)
(21% aq. soln.) clear, colorless
103.0°C to 104.0°C
98%
Authentic
MFCD00002853
100g
Solubility in water: soluble. Other solubilities: soluble in alcohol
C(CO)N(CCO)C(CO)(CO)CO
209.24
CHEBI:41250
Powder or Crystalline Powder
0.1% max.
97.5
100.0
White to Brown
0.2% max. (80°C, 5 hrs)
C8H19NO5
(HOCH2CH2)2NC(CH2OH)3
Plastic bottle
1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis 2-hydroxyethyl iminotris hydroxymethyl methane, bis-tris, bis-tris buffer, bistris, unii-q1xc3631cp
OWMVSZAMULFTJU-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
81462
209.24
98%
2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, 98%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 230-237-7

TSCA : TSCA

SDS
Product Certifications

RUO – Research Use Only