missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC163825000
Specifications
(0.1M in H2O) | |
2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol | |
0.2% max. (80°C, 5 hrs) | |
0.1% max. | |
103.0°C to 104.0°C | |
98% | |
97.5 | |
98% | |
(HOCH2CH2)2NC(CH2OH)3 | |
1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis 2-hydroxyethyl iminotris hydroxymethyl methane, bis-tris, bis-tris buffer, bistris, unii-q1xc3631cp | |
OWMVSZAMULFTJU-UHFFFAOYSA-N | |
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol | |
81462 | |
209.24 | |
98% |
(21% aq. soln.) clear, colorless | |
Authentic | |
Plastic bottle | |
White to Brown | |
500g | |
6976-37-0 | |
100.0 | |
C8H19NO5 | |
MFCD00002853 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
C(CO)N(CCO)C(CO)(CO)CO | |
209.24 | |
CHEBI:41250 | |
Powder or Crystalline Powder |
Chemical Identifiers
6976-37-0 | |
209.24 | |
OWMVSZAMULFTJU-UHFFFAOYSA-N | |
81462 | |
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol |
C8H19NO5 | |
MFCD00002853 | |
1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis 2-hydroxyethyl iminotris hydroxymethyl methane, bis-tris, bis-tris buffer, bistris, unii-q1xc3631cp | |
CHEBI:41250 | |
C(CO)N(CCO)C(CO)(CO)CO |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 230-237-7
TSCA : TSCA