Please login to your online account to display your discounted pricing

2,2-Bis(hydroxymethyl)-2,2',2'-nitrilotriethanol, 98%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  163821000

 View more versions of this product

Catalog No. AC163821000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (total base)
Physical Form Powder or Crystalline Powder
Formula Weight 209.24
Quantity 100g
Color White to brown
EINECS Number 230-237-7
TSCA TSCA
CAS Max % 100.0
Appearance of Solution (21% aq. soln.) clear, colorless
Chemical Name or Material 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol
Infrared Spectrum Authentic
Loss on Drying 0.2% max. (80°C, 5 hrs)
Name Note 98%
Molecular Formula C8H19NO5
Absorbance Conditions (0.1M in H2O)
Synonym BIS-TRIS
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Assay Percent Range 98%
Linear Formula (HOCH2CH2)2NC(CH2OH)3
MDL Number MFCD00002853
Packaging Plastic bottle
CAS 6976-37-0
Heavy Metals 5ppm max.
Absorbance 0.04 max. at 300nm
Iron (Fe) 5ppm max.
Sulfated Ash 0.1% max.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep only in original container.
Absorb spillage to prevent material damage.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May be corrosive to metals.

Specifications

Chemical Name or Material 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol
Name Note 98%
CAS 6976-37-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (total base)
Assay Percent Range 98%
Molecular Formula C8H19NO5
Linear Formula (HOCH2CH2)2NC(CH2OH)3
Formula Weight 209.24
MDL Number MFCD00002853
Synonym BIS-TRIS
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep only in original container.
Absorb spillage to prevent material damage.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May be corrosive to metals.

SDS & Safety Documents