missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC368640010

 View more versions of this product

Catalog No. AC368640010



Description

Description

Specifications

Specifications

(±)-2, 2'-Bis(diphenylphosphino)-1, 1'-binaphthyl
97.5% min.
White to Beige
1g
97.5
98%
((C6H5)2PC10H2)2
15,1227
MUALRAIOVNYAIW-UHFFFAOYSA-N
[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane
634876
Powder
Authentic
Glass bottle
280.0°C to 285.0°C
98327-87-8
100.0
C44H32P2
MFCD00010805
2,2'-bis diphenylphosphino-1,1'-binaphthalene, 2,2'-bis diphenylphosphino-1,1'-binaphthyl, binap, r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl, r-+-binap, r-binap, rac-binap, s---2,2'-bis diphenylphosphino-1,1'-binaphthyl, s---binap, s-binap
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8
622.69
622.69
98%

Chemical Identifiers

98327-87-8
622.69
MUALRAIOVNYAIW-UHFFFAOYSA-N
634876
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8
C44H32P2
MFCD00010805
2,2'-bis diphenylphosphino-1,1'-binaphthalene, 2,2'-bis diphenylphosphino-1,1'-binaphthyl, binap, r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl, r-+-binap, r-binap, rac-binap, s---2,2'-bis diphenylphosphino-1,1'-binaphthyl, s---binap, s-binap
[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents