Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, 98%

$67.58 - $612.64

Chemical Identifiers

Chemical Name or Material (+/-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl
CAS 153305-67-0
Molecular Formula C48H40P2
Formula Weight 678.79
MDL Number MFCD01311709
Synonym rac-p-Tol-BINAP
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2758503

  SDS
alfa aesar™
H2758503
1g Each for $166.24

AAH2758506

  SDS
alfa aesar™
H2758506
5g Each for $612.64

AAH27585MD

  SDS
alfa aesar™
H27585MD
250mg Each for $67.58
Description & Specifications

Specifications

Formula Weight 678.79
Chemical Name or Material (+/-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl
Assay 98%
Molecular Formula C48H40P2
MDL Number MFCD01311709
CAS 153305-67-0
Synonym rac-p-Tol-BINAP
Melting Point 267° to 271°C
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.