missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Bis(allyloxymethyl)-1-butanol (contains Mono- and Tri-substituted Product), TCI America™

$35.14 - $126.81

Chemical Identifiers

CAS 682-09-7
Molecular Formula C12H22O3
Molecular Weight (g/mol) 214.305
MDL Number MFCD00035791
InChI Key BDKDHWOPFRTWPP-UHFFFAOYSA-N
Synonym Trimethylolpropane Diallyl Ether
PubChem CID 61196
IUPAC Name 2,2-bis(prop-2-enoxymethyl)butan-1-ol
SMILES CCC(CO)(COCC=C)COCC=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B3003500G View Documents TCI America
B3003500G
500g Each for $126.81
 
B300325G View Documents TCI America
B300325G
25g Each for $35.14
 
Specifications

Chemical Identifiers

682-09-7
214.305
BDKDHWOPFRTWPP-UHFFFAOYSA-N
61196
CCC(CO)(COCC=C)COCC=C
C12H22O3
MFCD00035791
Trimethylolpropane Diallyl Ether
2,2-bis(prop-2-enoxymethyl)butan-1-ol

Specifications

122°C
500g
C12H22O3
Trimethylolpropane Diallyl Ether
CCC(CO)(COCC=C)COCC=C
214.305
214.31
≥85.0% (GC)
Yellow
682-09-7
MFCD00035791
BDKDHWOPFRTWPP-UHFFFAOYSA-N
2,2-bis(prop-2-enoxymethyl)butan-1-ol
61196
Clear Liquid at 20°C
2,2-Bis(allyloxymethyl)-1-butanol (contains Mono- and Tri-substituted Product)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0427

RTECSNumber : TY6667000

TSCA : Yes