Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Bis(4-methoxyphenyl)ethylamine, 98%

$74.58 - $263.69

Chemical Identifiers

Chemical Name or Material 2,2-Bis(4-methoxyphenyl)ethylamine
CAS 85336-83-0
Molecular Formula C16H19NO2
Formula Weight 257.34
MDL Number MFCD04039769
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2468903

  SDS
alfa aesar™
B2468903
1g Each for $74.58

AAB2468906

  SDS
alfa aesar™
B2468906
5g Each for $263.69
Description & Specifications

Specifications

Formula Weight 257.34
Chemical Name or Material 2,2-Bis(4-methoxyphenyl)ethylamine
Assay 98%
Molecular Formula C16H19NO2
MDL Number MFCD04039769
CAS 85336-83-0
Melting Point 45° to 50°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.