missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Bis(4-hydroxyphenyl)butane, TCI America™

$197.09 - $473.63

Chemical Identifiers

CAS 77-40-7
Molecular Formula C16H18O2
Molecular Weight (g/mol) 242.318
MDL Number MFCD00020179
InChI Key HTVITOHKHWFJKO-UHFFFAOYSA-N
Synonym bisphenol b, 2,2-bis 4-hydroxyphenyl butane, 4,4'-butane-2,2-diyl diphenol, phenol, 4,4'-1-methylpropylidene bis, p,p'-sec-butylidenediphenol, unii-1rc731tjja, 4,4'-1-methylpropylidene bisphenol, 1rc731tjja, 4,4'-sec-butylidenediphenol, 2,2-bis p-hydroxyphenyl butane
PubChem CID 66166
ChEBI CHEBI:34581
IUPAC Name 4-[2-(4-hydroxyphenyl)butan-2-yl]phenol
SMILES CCC(C)(C1=CC=C(C=C1)O)C2=CC=C(C=C2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B10505G View Documents TCI America
B10505G
5g Each for $197.09
 
B105025G View Documents TCI America
B105025G
25g Each for $473.63
 
Specifications

Chemical Identifiers

77-40-7
242.318
HTVITOHKHWFJKO-UHFFFAOYSA-N
66166
4-[2-(4-hydroxyphenyl)butan-2-yl]phenol
C16H18O2
MFCD00020179
bisphenol b, 2,2-bis 4-hydroxyphenyl butane, 4,4'-butane-2,2-diyl diphenol, phenol, 4,4'-1-methylpropylidene bis, p,p'-sec-butylidenediphenol, unii-1rc731tjja, 4,4'-1-methylpropylidene bisphenol, 1rc731tjja, 4,4'-sec-butylidenediphenol, 2,2-bis p-hydroxyphenyl butane
CHEBI:34581
CCC(C)(C1=CC=C(C=C1)O)C2=CC=C(C=C2)O

Specifications

White-Yellow
5g
C16H18O2
bisphenol b, 2,2-bis 4-hydroxyphenyl butane, 4,4'-butane-2,2-diyl diphenol, phenol, 4,4'-1-methylpropylidene bis, p,p'-sec-butylidenediphenol, unii-1rc731tjja, 4,4'-1-methylpropylidene bisphenol, 1rc731tjja, 4,4'-sec-butylidenediphenol, 2,2-bis p-hydroxyphenyl butane
CCC(C)(C1=CC=C(C=C1)O)C2=CC=C(C=C2)O
242.318
CHEBI:34581
Crystal-Powder at 20°C
2,2-Bis(4-hydroxyphenyl)butane
126°C
77-40-7
MFCD00020179
HTVITOHKHWFJKO-UHFFFAOYSA-N
4-[2-(4-hydroxyphenyl)butan-2-yl]phenol
66166
242.32
≥98.0% (GC)
Safety and Handling

Safety and Handling

RTECSNumber : SM1210200

TSCA : No