Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Bis(4-carboxyphenyl)hexafluoropropane, 95%

$29.55 - $83.43

Chemical Identifiers

Chemical Name or Material 2,2-Bis(4-carboxyphenyl)hexafluoropropane
CAS 1171-47-7
Molecular Formula C17H10F6O4
Formula Weight 392.26
MDL Number MFCD00040931
Beilstein 2022417
Synonym 4,4'-Hexafluoroisopropylidenebis(benzoic acid)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1045003

  SDS
alfa aesar™
L1045003
1g Each for $29.55

AAL1045006

  SDS
alfa aesar™
L1045006
5g Each for $83.43
Description & Specifications

Specifications

Formula Weight 392.26
Chemical Name or Material 2,2-Bis(4-carboxyphenyl)hexafluoropropane
MDL Number MFCD00040931
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 95%
Molecular Formula C17H10F6O4
Beilstein 2022417
Synonym 4,4'-Hexafluoroisopropylidenebis(benzoic acid)
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 1171-47-7
Melting Point 268° to 273°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.