missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Bis(4-carboxyphenyl)hexafluoropropane, 95%

Manufacturer:  Alfa Aesar™ L1045003

 View more versions of this product

Catalog No. AAL1045003



Specifications

Specifications

2,2-Bis(4-carboxyphenyl)hexafluoropropane
1g
C17H10F6O4
2022417
PHQYMDAUTAXXFZ-UHFFFAOYSA-N
4-[2-(4-carboxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]benzoic acid
633763
95%
268°C to 273°C
1171-47-7
MFCD00040931
2,2-bis 4-carboxyphenyl hexafluoropropane, 4,4'-perfluoropropane-2,2-diyl dibenzoic acid, benzoic acid, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis, 4-2-4-carboxyphenyl-1,1,1,3,3,3-hexafluoropropan-2-yl benzoic acid, 2,2-bis 4-carboxyphenyl hexafluoropropane, 4,4'-hexafluoroisopropylidene bis benzoic acid, 4-1-4-carboxyphenyl-2,2,2-trifluoro-1-trifluoromethyl ethyl benzoic acid, benzoic acid, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis, acmc-1c1gj, 2,2-bis-4-carboxyphenyl hexafluoropropane
C1=CC(=CC=C1C(=O)O)C(C2=CC=C(C=C2)C(=O)O)(C(F)(F)F)C(F)(F)F
392.253
392.26

Chemical Identifiers

1171-47-7
392.253
PHQYMDAUTAXXFZ-UHFFFAOYSA-N
633763
C1=CC(=CC=C1C(=O)O)C(C2=CC=C(C=C2)C(=O)O)(C(F)(F)F)C(F)(F)F
C17H10F6O4
MFCD00040931
2,2-bis 4-carboxyphenyl hexafluoropropane, 4,4'-perfluoropropane-2,2-diyl dibenzoic acid, benzoic acid, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis, 4-2-4-carboxyphenyl-1,1,1,3,3,3-hexafluoropropan-2-yl benzoic acid, 2,2-bis 4-carboxyphenyl hexafluoropropane, 4,4'-hexafluoroisopropylidene bis benzoic acid, 4-1-4-carboxyphenyl-2,2,2-trifluoro-1-trifluoromethyl ethyl benzoic acid, benzoic acid, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis, acmc-1c1gj, 2,2-bis-4-carboxyphenyl hexafluoropropane
4-[2-(4-carboxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents