missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Bis(4-allyloxy-3,5-dibromophenyl)propane, TCI America™

$47.36 - $233.75

Chemical Identifiers

CAS 25327-89-3
Molecular Formula C21H20Br4O2
Molecular Weight (g/mol) 624.005
MDL Number MFCD00086863
InChI Key PWXTUWQHMIFLKL-UHFFFAOYSA-N
Synonym Tetrabromobisphenol A Diallyl Ether
PubChem CID 61972
IUPAC Name 1,3-dibromo-5-[2-(3,5-dibromo-4-prop-2-enoxyphenyl)propan-2-yl]-2-prop-2-enoxybenzene
SMILES CC(C)(C1=CC(=C(C(=C1)Br)OCC=C)Br)C2=CC(=C(C(=C2)Br)OCC=C)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B202125G View Documents TCI America
B202125G
25g Each for $47.36
 
B2021500G View Documents TCI America
B2021500G
500g Each for $233.75
 
Specifications

Chemical Identifiers

25327-89-3
624.005
PWXTUWQHMIFLKL-UHFFFAOYSA-N
61972
CC(C)(C1=CC(=C(C(=C1)Br)OCC=C)Br)C2=CC(=C(C(=C2)Br)OCC=C)Br
C21H20Br4O2
MFCD00086863
Tetrabromobisphenol A Diallyl Ether
1,3-dibromo-5-[2-(3,5-dibromo-4-prop-2-enoxyphenyl)propan-2-yl]-2-prop-2-enoxybenzene

Specifications

White-Yellow
25g
C21H20Br4O2
Tetrabromobisphenol A Diallyl Ether
CC(C)(C1=CC(=C(C(=C1)Br)OCC=C)Br)C2=CC(=C(C(=C2)Br)OCC=C)Br
624.005
624.01
≥99.0% (HPLC)
120°C
25327-89-3
MFCD00086863
PWXTUWQHMIFLKL-UHFFFAOYSA-N
1,3-dibromo-5-[2-(3,5-dibromo-4-prop-2-enoxyphenyl)propan-2-yl]-2-prop-2-enoxybenzene
61972
Crystal-Powder at 20°C
2,2-Bis(4-allyloxy-3,5-dibromophenyl)propane
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0218

RTECSNumber : DA6126250

TSCA : Yes