missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Bis[4-(4-aminophenoxy)phenyl]propane, 98%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5505714

 View more versions of this product

Catalog No. AAH5505714



Description

Description

Applications
It is applied in pharmaceutical industry.

Solubility
Very slightly soluble in water. Soluble in acetone, Dimethyl sulfoxide(DMSO).

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
Specifications

Specifications

2,2-Bis[4-(4-aminophenoxy)phenyl]propane
25g
C27H26N2O2
2,2'-bis 4-aminophenoxyphenyl propane, 2,2-bis 4-4-aminophenoxy phenyl propane, 4,4'-1-methylethylidene bis 4,1-phenyleneoxy bisbenzenamine, 4,4'-4,4'-isopropylidenediphenyl-1,1'-diyldioxy dianiline, 4,4'-isopropylidenebis 4,1-phenyleneoxy dianiline, 4,4'-propane-2,2-diylbis 4,1-phenylene bis oxy dianiline, benzenamine, 4,4'-1-methylethylidene bis 4,1-phenyleneoxy bis, bis 4-4-aminophenoxy phenyl dimethyl methane
KMKWGXGSGPYISJ-UHFFFAOYSA-N
4-[4-[2-[4-(4-aminophenoxy)phenyl]propan-2-yl]phenoxy]aniline
83119
98%
127°C to 130°C
13080-86-9
MFCD00039152
Very slightly soluble in water. Soluble in acetone,Dimethyl sulfoxide(DMSO).
CC(C)(C1=CC=C(C=C1)OC2=CC=C(C=C2)N)C3=CC=C(C=C3)OC4=CC=C(C=C4)N
410.517
410.52

Chemical Identifiers

13080-86-9
410.517
KMKWGXGSGPYISJ-UHFFFAOYSA-N
83119
CC(C)(C1=CC=C(C=C1)OC2=CC=C(C=C2)N)C3=CC=C(C=C3)OC4=CC=C(C=C4)N
C27H26N2O2
MFCD00039152
2,2'-bis 4-aminophenoxyphenyl propane, 2,2-bis 4-4-aminophenoxy phenyl propane, 4,4'-1-methylethylidene bis 4,1-phenyleneoxy bisbenzenamine, 4,4'-4,4'-isopropylidenediphenyl-1,1'-diyldioxy dianiline, 4,4'-isopropylidenebis 4,1-phenyleneoxy dianiline, 4,4'-propane-2,2-diylbis 4,1-phenylene bis oxy dianiline, benzenamine, 4,4'-1-methylethylidene bis 4,1-phenyleneoxy bis, bis 4-4-aminophenoxy phenyl dimethyl methane
4-[4-[2-[4-(4-aminophenoxy)phenyl]propan-2-yl]phenoxy]aniline
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c

H302-H317

EINECSNumber : 235-985-8

RTECSNumber : CY1062500

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents