missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Bis(3-nitrophenyl)hexafluoropropane, TCI America™

$229.17 - $768.48

Chemical Identifiers

CAS 64465-34-5
Molecular Formula C15H8F6N2O4
Molecular Weight (g/mol) 394.229
MDL Number MFCD02093448
InChI Key YETXOTPYXAYEMI-UHFFFAOYSA-N
Synonym 3,3′-(Hexafluoroisopropylidene)bis(nitrobenzene)
PubChem CID 14029053
IUPAC Name 1-[1,1,1,3,3,3-hexafluoro-2-(3-nitrophenyl)propan-2-yl]-3-nitrobenzene
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(C2=CC(=CC=C2)[N+](=O)[O-])(C(F)(F)F)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B21151G View Documents TCI America
B21151G
1g Each for $229.17
 
B21155G View Documents TCI America
B21155G
5g Each for $768.48
 
Specifications

Chemical Identifiers

64465-34-5
394.229
YETXOTPYXAYEMI-UHFFFAOYSA-N
14029053
C1=CC(=CC(=C1)[N+](=O)[O-])C(C2=CC(=CC=C2)[N+](=O)[O-])(C(F)(F)F)C(F)(F)F
C15H8F6N2O4
MFCD02093448
3,3′-(Hexafluoroisopropylidene)bis(nitrobenzene)
1-[1,1,1,3,3,3-hexafluoro-2-(3-nitrophenyl)propan-2-yl]-3-nitrobenzene

Specifications

White-Yellow
1g
C15H8F6N2O4
3,3′C-(Hexafluoroisopropylidene)bis(nitrobenzene)
C1=CC(=CC(=C1)[N+](=O)[O-])C(C2=CC(=CC=C2)[N+](=O)[O-])(C(F)(F)F)C(F)(F)F
394.229
394.23
≥99.0% (T)
120°C
64465-34-5
MFCD02093448
YETXOTPYXAYEMI-UHFFFAOYSA-N
1-[1,1,1,3,3,3-hexafluoro-2-(3-nitrophenyl)propan-2-yl]-3-nitrobenzene
14029053
Crystal-Powder at 20°C
2,2-Bis(3-nitrophenyl)hexafluoropropane
Safety and Handling

Safety and Handling

TSCA : No