missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Bis(3-aminophenyl)hexafluoropropane, TCI America™

$721.13 - $2,276.44

Chemical Identifiers

CAS 47250-53-3
Molecular Formula C15H12F6N2
Molecular Weight (g/mol) 334.265
MDL Number MFCD00191619
InChI Key UVUCUHVQYAPMEU-UHFFFAOYSA-N
Synonym 3,3′-(Hexafluoroisopropylidene)dianiline
PubChem CID 630566
IUPAC Name 3-[2-(3-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]aniline
SMILES C1=CC(=CC(=C1)N)C(C2=CC(=CC=C2)N)(C(F)(F)F)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B170425G View Documents TCI America
B170425G
25g Each for $2,276.44
 
B17045G View Documents TCI America
B17045G
5g Each for $721.13
 
Specifications

Chemical Identifiers

47250-53-3
334.265
UVUCUHVQYAPMEU-UHFFFAOYSA-N
630566
C1=CC(=CC(=C1)N)C(C2=CC(=CC=C2)N)(C(F)(F)F)C(F)(F)F
C15H12F6N2
MFCD00191619
3,3′-(Hexafluoroisopropylidene)dianiline
3-[2-(3-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]aniline

Specifications

Undesignated
25g
C15H12F6N2
3,3′-(Hexafluoroisopropylidene)dianiline
C1=CC(=CC(=C1)N)C(C2=CC(=CC=C2)N)(C(F)(F)F)C(F)(F)F
334.265
334.27
≥98.0% (GC,T)
86°C
47250-53-3
MFCD00191619
UVUCUHVQYAPMEU-UHFFFAOYSA-N
3-[2-(3-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]aniline
630566
Crystal-Powder at 20°C
2,2-Bis(3-aminophenyl)hexafluoropropane
Safety and Handling

Safety and Handling

TSCA : No