missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Bis(3-amino-4-methylphenyl)hexafluoropropane, TCI America™

$110.00 - $328.48

Chemical Identifiers

CAS 116325-74-7
Molecular Formula C17H16F6N2
Molecular Weight (g/mol) 362.319
MDL Number MFCD00039148
InChI Key HJSYPLCSZPEDCQ-UHFFFAOYSA-N
Synonym 5,5′-(Hexafluoroisopropylidene)di-o-toluidine
PubChem CID 632384
IUPAC Name 5-[2-(3-amino-4-methylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-2-methylaniline
SMILES CC1=C(C=C(C=C1)C(C2=CC(=C(C=C2)C)N)(C(F)(F)F)C(F)(F)F)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B13975G View Documents TCI America
B13975G
5g Each for $110.00
 
B139725G View Documents TCI America
B139725G
25g Each for $328.48
 
Specifications

Chemical Identifiers

116325-74-7
362.319
HJSYPLCSZPEDCQ-UHFFFAOYSA-N
632384
CC1=C(C=C(C=C1)C(C2=CC(=C(C=C2)C)N)(C(F)(F)F)C(F)(F)F)N
C17H16F6N2
MFCD00039148
5,5′-(Hexafluoroisopropylidene)di-o-toluidine
5-[2-(3-amino-4-methylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-2-methylaniline

Specifications

Undesignated
5g
C17H16F6N2
5,5′C-(Hexafluoroisopropylidene)di-o-toluidine
CC1=C(C=C(C=C1)C(C2=CC(=C(C=C2)C)N)(C(F)(F)F)C(F)(F)F)N
362.319
362.32
≥98.0% (HPLC,T)
115°C
116325-74-7
MFCD00039148
HJSYPLCSZPEDCQ-UHFFFAOYSA-N
5-[2-(3-amino-4-methylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-2-methylaniline
632384
Crystal-Powder at 20°C
2,2-Bis(3-amino-4-methylphenyl)hexafluoropropane
Safety and Handling

Safety and Handling

TSCA : No