missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane, TCI America™

$91.67 - $270.42

Chemical Identifiers

CAS 83558-87-6
Molecular Formula C15H12F6N2O2
Molecular Weight (g/mol) 366.263
MDL Number MFCD00039147
InChI Key MSTZGVRUOMBULC-UHFFFAOYSA-N
Synonym 2,2-bis 3-amino-4-hydroxyphenyl hexafluoropropane, 4,4'-perfluoropropane-2,2-diyl bis 2-aminophenol, 2,2-bis 3-amino-4-hydroxyphenyl-hexafluoropropane, 2-amino-4-2-3-amino-4-hydroxyphenyl-1,1,1,3,3,3-hexafluoropropan-2-yl phenol, 4,4'-hexafluoroisopropylidene bis 2-aminophenol, phenol, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis 2-amino, 4,4'-1,1,1,3,3,3-hexafluoropropane-2,2-diyl bis 2-aminophenol, 2,2-bis 3-amino-4-hydrophenyl hexafluoropropane, 1,1-bis 3-amino-4-hydroxyphenyl-2,2,2-trifluoro-1-trifluoromethyl ethane, phenol, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis 2-amino
PubChem CID 632603
IUPAC Name 2-amino-4-[2-(3-amino-4-hydroxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]phenol
SMILES C1=CC(=C(C=C1C(C2=CC(=C(C=C2)O)N)(C(F)(F)F)C(F)(F)F)N)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B14155G View Documents TCI America
B14155G
5g Each for $91.67
 
B141525G View Documents TCI America
B141525G
25g Each for $270.42
 
Specifications

Chemical Identifiers

83558-87-6
366.263
MSTZGVRUOMBULC-UHFFFAOYSA-N
632603
C1=CC(=C(C=C1C(C2=CC(=C(C=C2)O)N)(C(F)(F)F)C(F)(F)F)N)O
C15H12F6N2O2
MFCD00039147
2,2-bis 3-amino-4-hydroxyphenyl hexafluoropropane, 4,4'-perfluoropropane-2,2-diyl bis 2-aminophenol, 2,2-bis 3-amino-4-hydroxyphenyl-hexafluoropropane, 2-amino-4-2-3-amino-4-hydroxyphenyl-1,1,1,3,3,3-hexafluoropropan-2-yl phenol, 4,4'-hexafluoroisopropylidene bis 2-aminophenol, phenol, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis 2-amino, 4,4'-1,1,1,3,3,3-hexafluoropropane-2,2-diyl bis 2-aminophenol, 2,2-bis 3-amino-4-hydrophenyl hexafluoropropane, 1,1-bis 3-amino-4-hydroxyphenyl-2,2,2-trifluoro-1-trifluoromethyl ethane, phenol, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis 2-amino
2-amino-4-[2-(3-amino-4-hydroxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]phenol

Specifications

White
5g
C15H12F6N2O2
2,2-bis 3-amino-4-hydroxyphenyl hexafluoropropane, 4,4'-perfluoropropane-2,2-diyl bis 2-aminophenol, 2,2-bis 3-amino-4-hydroxyphenyl-hexafluoropropane, 2-amino-4-2-3-amino-4-hydroxyphenyl-1,1,1,3,3,3-hexafluoropropan-2-yl phenol, 4,4'-hexafluoroisopropylidene bis 2-aminophenol, phenol, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis 2-amino, 4,4'-1,1,1,3,3,3-hexafluoropropane-2,2-diyl bis 2-aminophenol, 2,2-bis 3-amino-4-hydrophenyl hexafluoropropane, 1,1-bis 3-amino-4-hydroxyphenyl-2,2,2-trifluoro-1-trifluoromethyl ethane, phenol, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis 2-amino
C1=CC(=C(C=C1C(C2=CC(=C(C=C2)O)N)(C(F)(F)F)C(F)(F)F)N)O
366.263
366.26
≥98.0% (T)
243°C
83558-87-6
MFCD00039147
MSTZGVRUOMBULC-UHFFFAOYSA-N
2-amino-4-[2-(3-amino-4-hydroxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]phenol
632603
Crystal-Powder at 20°C
2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1804

TSCA : Yes