missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Bis(3,4-dimethylphenyl)hexafluoropropane, TCI America™

$134.45 - $165.00

Chemical Identifiers

CAS 65294-20-4
Molecular Formula C19H18F6
Molecular Weight (g/mol) 360.343
MDL Number MFCD00042167
InChI Key GLFKFHJEFMLTOB-UHFFFAOYSA-N
Synonym 4,4′C-(Hexafluoroisopropylidene)di-o-xylene
PubChem CID 103356
IUPAC Name 4-[2-(3,4-dimethylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-1,2-dimethylbenzene
SMILES CC1=C(C=C(C=C1)C(C2=CC(=C(C=C2)C)C)(C(F)(F)F)C(F)(F)F)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B140210G View Documents TCI America
B140210G
10g Each for $165.00
 
B14025G View Documents TCI America
B14025G
5g Each for $134.45
 
Specifications

Chemical Identifiers

65294-20-4
360.343
GLFKFHJEFMLTOB-UHFFFAOYSA-N
103356
CC1=C(C=C(C=C1)C(C2=CC(=C(C=C2)C)C)(C(F)(F)F)C(F)(F)F)C
C19H18F6
MFCD00042167
4,4′C-(Hexafluoroisopropylidene)di-o-xylene
4-[2-(3,4-dimethylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-1,2-dimethylbenzene

Specifications

White
10g
C19H18F6
4,4′C-(Hexafluoroisopropylidene)di-o-xylene
CC1=C(C=C(C=C1)C(C2=CC(=C(C=C2)C)C)(C(F)(F)F)C(F)(F)F)C
360.343
360.34
≥97.0% (GC)
78°C
65294-20-4
MFCD00042167
GLFKFHJEFMLTOB-UHFFFAOYSA-N
4-[2-(3,4-dimethylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-1,2-dimethylbenzene
103356
Crystal-Powder at 20°C
2,2-Bis(3,4-dimethylphenyl)hexafluoropropane
Safety and Handling

Safety and Handling

TSCA : Yes