missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2'-Bipyrimidine, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L01190MD
Description
2,2'-Bipyrimidine is used in the preparation of 5-bromo-[2,2']bipyrimidinyl.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2′-Bipyrimidine is used in the preparation of 5-bromo-[2,2′]bipyrimidinyl.
Solubility
Soluble in methanol.
Notes
Incompatible with strong oxidizing agents.
Specifications
2,2'-Bipyrimidine | |
110°C to 116°C | |
MFCD00014600 | |
607396 | |
Soluble in methanol. | |
C1=CN=C(N=C1)C1=NC=CC=N1 | |
158.16 | |
158.16 |
34671-83-5 | |
C8H6N4 | |
250 mg | |
2,2'-bipyrimidine, 2,2'-bipyrimidyl, 2,2-bipyrimidine, bpym, 2-pyrimidin-2-yl pyrimidine, bipyrimidine, 2,2'-bipyrimidinyl, 2,2'-dipyrimidyl, 2,2;-bipyrimidine | |
HKOAFLAGUQUJQG-UHFFFAOYSA-N | |
2-pyrimidin-2-ylpyrimidine | |
123444 | |
96% |
Chemical Identifiers
34671-83-5 | |
158.16 | |
HKOAFLAGUQUJQG-UHFFFAOYSA-N | |
123444 |
C8H6N4 | |
MFCD00014600 | |
2,2'-bipyrimidine, 2,2'-bipyrimidyl, 2,2-bipyrimidine, bpym, 2-pyrimidin-2-yl pyrimidine, bipyrimidine, 2,2'-bipyrimidinyl, 2,2'-dipyrimidyl, 2,2;-bipyrimidine | |
C1=CN=C(N=C1)C1=NC=CC=N1 |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 252-137-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only