missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ 2,2′-Bipyridyl, ≥99.0% (NT), Honeywell™ Fluka™

Redox indicator, for spectrophotometric det. of Fe,≥99.0%

$63.55 - $543.81

Chemical Identifiers

CAS 366-18-7
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.188
MDL Number MFCD00006212
InChI Key ROFVEXUMMXZLPA-UHFFFAOYSA-N
PubChem CID 1474
ChEBI CHEBI:30351
IUPAC Name 2-pyridin-2-ylpyridine
SMILES C1=CC=NC(=C1)C2=CC=CC=N2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60044860 View Documents Honeywell Fluka™
14454100G
100g Glass Bottle Each for $543.81
 
60044861 View Documents Honeywell Fluka™
1445425G
25g Plastic Bottle Each for $216.89
 
60044862 View Documents Honeywell Fluka™
144545G
5g Glass Bottle Each for $63.55
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

366-18-7
156.188
ROFVEXUMMXZLPA-UHFFFAOYSA-N
CHEBI:30351
C1=CC=NC(=C1)C2=CC=CC=N2
C10H8N2
MFCD00006212
1474
2-pyridin-2-ylpyridine

Specifications

Redox indicator, for spectrophotometric determination of Fe
C10H8N2
UN2811
≤0.05%
C1=CC=NC(=C1)C2=CC=CC=N2
156.188
CHEBI:30351
≥99.0% (NT)
Solid
69°C to 71°C
2,2′-Bipyridyl
366-18-7
MFCD00006212
113089
ROFVEXUMMXZLPA-UHFFFAOYSA-N
2-pyridin-2-ylpyridine
1474
156.18g/mol
Glass Bottle
273°C
100g
Safety and Handling

Safety and Handling

P280-P301 + P310-P312

H301 + H311

DOTInformation : 6.1/PGIII

EINECSNumber : 206-674-4

RTECSNumber : DW1750000