missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2′-Bipyridyl, ≥99.0% (NT), Honeywell Fluka™

Redox indicator, for spectrophotometric det. of Fe,≥99.0%

Manufacturer:  Honeywell-Fluka 14454100G

 View more versions of this product

Catalog No. 60044860



Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

2,2′-Bipyridyl
366-18-7
MFCD00006212
113089
ROFVEXUMMXZLPA-UHFFFAOYSA-N
2-pyridin-2-ylpyridine
1474
156.18g/mol
Glass Bottle
273°C
100g
Redox indicator, for spectrophotometric determination of Fe
C10H8N2
UN2811
≤0.05%
C1=CC=C(N=C1)C1=CC=CC=N1
156.19
CHEBI:30351
≥99.0% (NT)
Solid
69°C to 71°C

Chemical Identifiers

366-18-7
156.19
ROFVEXUMMXZLPA-UHFFFAOYSA-N
CHEBI:30351
C1=CC=C(N=C1)C1=CC=CC=N1
C10H8N2
MFCD00006212
1474
2-pyridin-2-ylpyridine
Safety and Handling

Safety and Handling

P280-P301 + P310-P312

H301 + H311

DOTInformation : 6.1/PGIII

EINECSNumber : 206-674-4

RTECSNumber : DW1750000

SDS
Documents
Product Certifications