missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2'-Bipyridine, ACS, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar 3056930

 View more versions of this product

Catalog No. AA3056930



Specifications

Specifications

2,2'-Bipyridine
272°C to 273°C
70°C
366-18-7
C10H8N2
UN2811
14,3347
ROFVEXUMMXZLPA-UHFFFAOYSA-N
2-pyridin-2-ylpyridine
1474
156.19
98%
121° (249°F)
White
250g
ACS
MFCD00006212
113089
2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl
C1=CC=NC(=C1)C2=CC=CC=N2
156.188
CHEBI:30351
Crystalline

Chemical Identifiers

366-18-7
156.188
ROFVEXUMMXZLPA-UHFFFAOYSA-N
1474
2-pyridin-2-ylpyridine
C10H8N2
MFCD00006212
2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl
CHEBI:30351
C1=CC=NC(=C1)C2=CC=CC=N2
Safety and Handling

Safety and Handling

GHS H Statement
H301-H312
Toxic if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P310-P312-P363-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : DW1750000

TSCA : TSCA

SDS Documents