missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2′-Bipyridine-6-carbonitrile, TCI America™

$273.48 - $951.83

Chemical Identifiers

CAS 4392-85-2
Molecular Formula C11H7N3
Molecular Weight (g/mol) 181.198
MDL Number MFCD07364147
InChI Key QLZHLNAALBJVOE-UHFFFAOYSA-N
Synonym 6-Cyano-2,2′C-bipyridyl, 6-Cyano-2,2′C-bipyridine
PubChem CID 11095236
IUPAC Name 6-pyridin-2-ylpyridine-2-carbonitrile
SMILES C1=CC=NC(=C1)C2=CC=CC(=N2)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B4557200MG View Documents TCI America
B4557200MG
200mg Each for $273.48
 
B45571G View Documents TCI America
B45571G
1g Each for $951.83
 
Specifications

Chemical Identifiers

4392-85-2
181.198
QLZHLNAALBJVOE-UHFFFAOYSA-N
11095236
C1=CC=NC(=C1)C2=CC=CC(=N2)C#N
C11H7N3
MFCD07364147
6-Cyano-2,2′C-bipyridyl, 6-Cyano-2,2′C-bipyridine
6-pyridin-2-ylpyridine-2-carbonitrile

Specifications

Undesignated
200mg
C11H7N3
6-Cyano-2,2′C-bipyridyl, 6-Cyano-2,2′C-bipyridine
C1=CC=NC(=C1)C2=CC=CC(=N2)C#N
181.198
181.20
≥98.0% (GC)
133°C
4392-85-2
MFCD07364147
QLZHLNAALBJVOE-UHFFFAOYSA-N
6-pyridin-2-ylpyridine-2-carbonitrile
11095236
Crystal-Powder at 20°C
2,2′-Bipyridine-6-carbonitrile
Safety and Handling

Safety and Handling

TSCA : No