missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2′-Bipyridine-3,3′-diol, TCI America™

$215.42 - $753.20

Chemical Identifiers

CAS 36145-03-6
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD00075177
InChI Key ZPHMTLAGULLPGM-KTKRTIGZSA-N
Synonym 3,3′C-Dihydroxy-2,2′C-bipyridyl, 3,3′C-Dihydroxy-2,2′C-bipyridine
PubChem CID 16212186
IUPAC Name (2Z)-2-(3-hydroxy-1H-pyridin-2-ylidene)pyridin-3-one
SMILES C1=CC(=O)C(=C2C(=CC=CN2)O)N=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B425550MG View Documents TCI America
B425550MG
50mg Each for $215.42
 
B4255250MG View Documents TCI America
B4255250MG
250mg Each for $753.20
 
Specifications

Chemical Identifiers

36145-03-6
188.186
ZPHMTLAGULLPGM-KTKRTIGZSA-N
16212186
C1=CC(=O)C(=C2C(=CC=CN2)O)N=C1
C10H8N2O2
MFCD00075177
3,3′C-Dihydroxy-2,2′C-bipyridyl, 3,3′C-Dihydroxy-2,2′C-bipyridine
(2Z)-2-(3-hydroxy-1H-pyridin-2-ylidene)pyridin-3-one

Specifications

Undesignated
50mg
C10H8N2O2
3,3′C-Dihydroxy-2,2′C-bipyridyl, 3,3′C-Dihydroxy-2,2′C-bipyridine
C1=CC(=O)C(=C2C(=CC=CN2)O)N=C1
188.186
188.19
≥98.0% (GC)
199°C
36145-03-6
MFCD00075177
ZPHMTLAGULLPGM-KTKRTIGZSA-N
(2Z)-2-(3-hydroxy-1H-pyridin-2-ylidene)pyridin-3-one
16212186
Crystal-Powder at 20°C
2,2′-Bipyridine-3,3′-diol
Safety and Handling

Safety and Handling

TSCA : No