Please login to your online account to display your discounted pricing

2,2'-bicinchoninic acid disodium salt, MP Biomedicals™


Quantity 5g

Manufacturer: mp biomedicals  15241405

 View more versions of this product

Catalog No. ICN15241405

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 388.3
Physical Form Powder
Color White
Quantity 5g
Chemical Name or Material 2, 2'-bicinchoninic acid disodium salt
CAS 979-88-4
Molecular Formula C20H10N2O4Na2
Synonym 4, 4'-Dicarboxy-2, 2-biquinoline disodium salt
Recommended Storage Store at Room Temperature (15° to 30°C).

Specifications

Chemical Name or Material 2, 2'-bicinchoninic acid disodium salt
CAS 979-88-4
Physical Form Powder
Molecular Formula C20H10N2O4Na2
Formula Weight 388.3
Synonym 4, 4'-Dicarboxy-2, 2-biquinoline disodium salt
Accessories

Store at Room Temperature (15° to 30°C).