Please login to your online account to display your discounted pricing

2,2'-bicinchoninic acid disodium salt, MP Biomedicals™

$258.27 - $258.27

Chemical Identifiers

Chemical Name or Material 2, 2'-bicinchoninic acid disodium salt
CAS 979-88-4
Physical Form Powder
Molecular Formula C20H10N2O4Na2
Formula Weight 388.3
Synonym 4, 4'-Dicarboxy-2, 2-biquinoline disodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15241405

 
mp biomedicals
15241405
5g Each for $258.27
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 388.3
Color White
Quantity 5g
Chemical Name or Material 2, 2'-bicinchoninic acid disodium salt
Molecular Formula C20H10N2O4Na2
CAS 979-88-4
Synonym 4, 4'-Dicarboxy-2, 2-biquinoline disodium salt
Recommended Storage Store at Room Temperature (15° to 30°C).
Safety and Handling

Store at Room Temperature (15° to 30°C).