Learn More
2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, 98%, Thermo Scientific Chemicals
Chromogenic substrate for peroxidase assays
$108.48 - $364.84
Chemical Identifiers
CAS | 30931-67-0 |
---|---|
Molecular Formula | C18H24N6O6S4 |
Molecular Weight (g/mol) | 548.67 |
MDL Number | MFCD00010404,MFCD00010404 |
InChI Key | OHDRQQURAXLVGJ-AXMZSLBLSA-N |
Synonym | ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate |
PubChem CID | 91884754 |
IUPAC Name | diazanium;3-ethyl-2-[(E)-(3-ethyl-6-sulfonato-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonate |
SMILES | [NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6553503
|
Thermo Scientific Chemicals
J6553503 |
1 g |
Each for $108.48
|
|
|||||
AAJ6553506
|
Thermo Scientific Chemicals
J6553506 |
5 g |
Each for $364.84
|
|
|||||
Description
Chromogenic substrate for peroxidase assays.2,2'-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt is widely utilized as a peroxidase substrate suitable for use in enzyme-linked immunosorbent assay (ELISA) procedures. It plays an important role in the food industry to measure the antioxidant capacities of foods. It is used to observe the reaction kinetics of specific enzymes and involved in glucose estimating reagent in blood serum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChromogenic substrate for peroxidase assays.2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt is widely utilized as a peroxidase substrate suitable for use in enzyme-linked immunosorbent assay (ELISA) procedures. It plays an important role in the food industry to measure the antioxidant capacities of foods. It is used to observe the reaction kinetics of specific enzymes and involved in glucose estimating reagent in blood serum.
Solubility
Dissolve in water at 50mg/ml. Solution may be slightly hazy.
Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
30931-67-0 | |
548.67 | |
OHDRQQURAXLVGJ-AXMZSLBLSA-N | |
91884754 | |
[NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O |
C18H24N6O6S4 | |
MFCD00010404,MFCD00010404 | |
ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate | |
diazanium;3-ethyl-2-[(E)-(3-ethyl-6-sulfonato-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonate |
Specifications
30931-67-0 | |
C18H24N6O6S4 | |
1 g | |
Light sensitive | |
Dissolve in water at 50mg/ml. Solution may be slightly hazy. | |
[NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O | |
548.67 | |
548.68 | |
Powder |
Green | |
MFCD00010404,MFCD00010404 | |
7329461 | |
ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate | |
OHDRQQURAXLVGJ-AXMZSLBLSA-N | |
diazanium;3-ethyl-2-[(E)-(3-ethyl-6-sulfonato-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonate | |
91884754 | |
98% | |
2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 250-396-6
RTECSNumber : DL7002000
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only