Please login to your online account to display your discounted pricing

2-(2-Aminoethyl)-1-methylpyrrolidine, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC160860050

 View more versions of this product

Catalog No. AC160860050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥96.0% (GC)
Formula Weight 128.22
Quantity 5g
Color Light beige to light brown
Density 0.8800g/mL
CAS Min % 96.0
CAS Max % 100.0
Refractive Index 1.4670 to 1.4690 (20°C, 589nm)
Assay Percent Range 97%
CAS 51387-90-7
Infrared Spectrum Authentic
Chemical Name or Material 2-(2-Aminoethyl)-1-methylpyrrolidine
Molecular Formula C7H16N2
MDL Number MFCD00003175
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. Harmful if swallowed.
Packaging Glass bottle
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

Specifications

Chemical Name or Material 2-(2-Aminoethyl)-1-methylpyrrolidine
CAS 51387-90-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C7H16N2
Formula Weight 128.22
MDL Number MFCD00003175
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

GHS P Statement Causes severe skin burns and eye damage. Harmful if swallowed.

SDS & Safety Documents